alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol
alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol
alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol
Thermo Scientific Chemicals

alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol

CAS: 1195-32-0 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H31739.06
also known as H31739-06
Price (EUR)
58,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
58,00
Each
Chemical Identifiers
CAS1195-32-0
IUPAC Name1-methyl-4-(prop-1-en-2-yl)benzene
Molecular FormulaC10H12
InChI KeyMMSLOZQEMPDGPI-UHFFFAOYSA-N
SMILESCC(=C)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Appearance (Color)Clear, colourless to pale yellow
Refractive Index1.5300-1.5380
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only