Thermo Scientific Chemicals

Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals

Catalog number: H31758.22
100 g, Each
Thermo Scientific Chemicals

Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals

Catalog number: H31758.22
100 g, Each
Quantity
Catalog number: H31758.22
also known as H31758-22
Price (USD)
Price: 1,051.00
Online price: 909.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4664-01-1
IUPAC Name
1H,2H,3H-pyrrolo[3,4-c]pyridine-1,3-dione
Molecular Formula
C7H4N2O2
InChI Key
SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CN=CC=C12
Melting Point (clear melt)
227.0-236.0?C
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Appearance (Color)
White to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text