Thermo Scientific Chemicals

4-Iodobutyl acetate, 96%, stab. with copper, Thermo Scientific Chemicals

Catalog number: H31826.18
50 g, Each
Thermo Scientific Chemicals

4-Iodobutyl acetate, 96%, stab. with copper, Thermo Scientific Chemicals

Catalog number: H31826.18
50 g, Each
Quantity
Catalog number: H31826.18
also known as H31826-18
Price (USD)
Price: 227.00
Online price: 196.65
Your price:
Quantity
-

Chemical Identifiers

CAS
40596-44-9
IUPAC Name
4-iodobutyl acetate
Molecular Formula
C6H11IO2
InChI Key
FLPVVTDEKLGZDZ-UHFFFAOYSA-N
SMILES
CC(=O)OCCCCI
Form
liquid
Assay (GC)
>96.0%
Appearance (Color)
Clear, yellow to brown or red/brown
Refractive Index
1.4985 - 1.5035 @ 20?C

Description

4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text