Specifications
CAS1047626-77-6
Chemical Name or Material2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole
Formula Weight278.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more