Thermo Scientific Chemicals

6-Bromochromone-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H31905.06
5 g, Each
Thermo Scientific Chemicals

6-Bromochromone-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H31905.06
5 g, Each
Quantity
Catalog number: H31905.06
also known as H31905-06
Price (USD)
Price: 422.00
Online price: 368.65
Your price:
Quantity
-

Chemical Identifiers

CAS
51085-91-7
IUPAC Name
6-bromo-4-oxo-4H-chromene-3-carboxylic acid
Molecular Formula
C10H5BrO4
InChI Key
IJTWNMVKNJSGMS-UHFFFAOYSA-N
SMILES
OC(=O)C1=COC2=CC=C(Br)C=C2C1=O
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥96.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text