2,4-Difluoro-5-nitrophenol, 98%
2,4-Difluoro-5-nitrophenol, 98%
2,4-Difluoro-5-nitrophenol, 98%
Thermo Scientific Chemicals

2,4-Difluoro-5-nitrophenol, 98%

CAS: 113512-57-5 | C6H3F2NO3 | 175.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32081.06
also known as H32081-06
Price (EUR)
90,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
90,00
Each
Chemical Identifiers
CAS113512-57-5
IUPAC Name2,4-difluoro-5-nitrophenol
Molecular FormulaC6H3F2NO3
InChI KeySMRYCTJAGPDVEH-UHFFFAOYSA-N
SMILESOC1=C(F)C=C(F)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)113.0-120.0?C
FormPowder
Appearance (Color)Cream to yellow
Identification (FTIR)Conforms
2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

Solubility
Sparingly soluble in water but readily soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only