4-(tert-Butyl)-o-phenylenediamine, 97%
4-(tert-Butyl)-o-phenylenediamine, 97%
4-(tert-Butyl)-o-phenylenediamine, 97%
Thermo Scientific Chemicals

4-(tert-Butyl)-o-phenylenediamine, 97%

CAS: 68176-57-8 | C10H16N2 | 164.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32205.03
also known as H32205-03
Price (EUR)
48,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
48,20
Each
Chemical Identifiers
CAS68176-57-8
IUPAC Name4-tert-butylbenzene-1,2-diamine
Molecular FormulaC10H16N2
InChI KeyWLOSFXSXVXTKBU-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(N)=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)94.0-104.0?C
Appearance (Color)Brown or purple
Assay (GC)≥96.0%
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

Solubility
Soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. M Strukelj.; F Papadimitrakopoulos.; TM Miller. Design and application of electron-transporting organic materials. Science. 1995267 (1), 5206-5209.
  2. A Maleki. Fe 3 O 4/SiO 2 nanoparticles: an efficient and magnetically recoverable nanocatalyst for the one-pot multicomponent synthesis of diazepines. Tetrahedron. 201268 (38), 7827-7833.