Dimethyl pyridine-2,6-dicarboxylate, 98%
Dimethyl pyridine-2,6-dicarboxylate, 98%
Dimethyl pyridine-2,6-dicarboxylate, 98%
Thermo Scientific Chemicals

Dimethyl pyridine-2,6-dicarboxylate, 98%

CAS: 5453-67-8 | C9H9NO4 | 195.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H32251.09
also known as H32251-09
Price (EUR)
48,30
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
48,30
Each
Chemical Identifiers
CAS5453-67-8
IUPAC Name2,6-dimethyl pyridine-2,6-dicarboxylate
Molecular FormulaC9H9NO4
InChI KeySNQQJEJPJMXYTR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=N1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)119.5-125.5?C
Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

Solubility
Soluble in water(14 g/L) (25°C). Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. J.-Y. Huang,l,; W. Xu. Dimethyl pyridine-2,6-dicarboxyl­ate . Acta Cryst. 200662 2653-2654.
  2. Mohammad Hakimia,; Keyvan Moeinia,; Zahra Mardania,; Esther Schuhb,; Fabian Mohrb. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies. Journal name. 2013, 66(7),1129-1141.