6-Methoxy-2-methylbenzothiazole, 97%
6-Methoxy-2-methylbenzothiazole, 97%
6-Methoxy-2-methylbenzothiazole, 97%
Thermo Scientific Chemicals

6-Methoxy-2-methylbenzothiazole, 97%

CAS: 2941-72-2 | C9H9NOS | 179.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32425.03
also known as H32425-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2941-72-2
IUPAC Name6-methoxy-2-methyl-1,3-benzothiazole
Molecular FormulaC9H9NOS
InChI KeyDYHLJSUORLPGNT-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=C(C)SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.6100-1.6150 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

General References:

  1. M Murray.; AJ Ryan. The binding to oxidised cytochromes P-450 and inhibition of mixed-function oxidases by aryl-substituted benzimidazoles and related compounds. Chemico-biological interactions.198343 (3), 341-351.
  2. WK Warburton. Arylthiazathiolium Salts And o-Aminoaryl Thiols-The Herz Reaction. Chemical Reviews.195757 (5), 1011-1020.