2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%
Thermo Scientific Chemicals

2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%

CAS: 852062-17-0 | C6H4F4N2 | 180.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32532.03
also known as H32532-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS852062-17-0
IUPAC Name3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Molecular FormulaC6H4F4N2
InChI KeyHAVVQRHBCUBQGG-UHFFFAOYSA-N
SMILESNC1=C(F)C=C(C=N1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)63.0-72.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only