3-Bromo-5-methoxybenzoic acid, 98%
3-Bromo-5-methoxybenzoic acid, 98%
3-Bromo-5-methoxybenzoic acid, 98%
Thermo Scientific Chemicals

3-Bromo-5-methoxybenzoic acid, 98%

CAS: 157893-14-6 | C8H7BrO3 | 231.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32635.03
also known as H32635-03
Price (EUR)
36,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
36,10
Each
Chemical Identifiers
CAS157893-14-6
IUPAC Name3-bromo-5-methoxybenzoic acid
Molecular FormulaC8H7BrO3
InChI KeyDMXJBCHYVUGXEH-UHFFFAOYSA-N
SMILESCOC1=CC(Br)=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)190.0-196.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only