3-Benzyloxybenzoic acid, 98%
3-Benzyloxybenzoic acid, 98%
3-Benzyloxybenzoic acid, 98%
Thermo Scientific Chemicals

3-Benzyloxybenzoic acid, 98%

CAS: 69026-14-8 | C14H11O3 | 227.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H32882.06
also known as H32882-06
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS69026-14-8
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to cream
Melting Point (clear melt)132.0-138.0?C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R. Amaranatha Reddy.; B. K. Sadashivai. Direct transition from a nematic phase to a polar biaxial smectic A phase in a homologous series of unsymmetrically substituted bent-core compounds. J. Mater. Chem. 2004, 14 (3), 310-319.
  2. B. K. Sadashiva.; R. Amaranatha Reddy.; R. Pratibha.; N. V. Madhusudana. Biaxial smectic A phase in homologous series of compounds composed of highly polar unsymmetrically substituted bent-core molecules. J. Mater. Chem. 2002, 12 (4), 943-950.