6-Bromo-2,3-difluorotoluene, 95%
6-Bromo-2,3-difluorotoluene, 95%
6-Bromo-2,3-difluorotoluene, 95%
Thermo Scientific Chemicals

6-Bromo-2,3-difluorotoluene, 95%

CAS: 847502-81-2 | C7H5BrF2 | 207.018 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32941.03
also known as H32941-03
Price (USD)
89.65
Online Exclusive
99.80
Save 10.15 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
89.65
Online Exclusive
99.80
Save 10.15 (10%)
Each
Chemical Identifiers
CAS847502-81-2
IUPAC Name1-bromo-3,4-difluoro-2-methylbenzene
Molecular FormulaC7H5BrF2
InChI KeyYXTCBTSPHSNDHD-UHFFFAOYSA-N
SMILESCC1=C(Br)C=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Refractive Index1.5065-1.5125 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only