Thermo Scientific Chemicals

5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific Chemicals

Catalog number: H33012.ME
500 mg, Each
Thermo Scientific Chemicals

5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific Chemicals

Catalog number: H33012.ME
500 mg, Each
Quantity
Catalog number: H33012.ME
also known as H33012-ME
Price (USD)
Quantity
-

Chemical Identifiers

CAS
500112-74-3
IUPAC Name
5-(4-methylphenyl)-2H,3H-[1,2,4]triazolo[3,4-b][1,3]thiazole-3-thione
Molecular Formula
C11H9N3S2
InChI Key
LNDYWRMGFKFDTC-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C1=CSC2=NNC(=S)N12
Form
Powder
Assay (HPLC)
≥95.0%
Appearance (Color)
Pale yellow
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
193.0-203.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text