4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
Thermo Scientific Chemicals

4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%

CAS: 705-24-8 | C5HCl2F3N2 | 216.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33142.MD
also known as H33142-MD
Price (EUR)
31,60
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
31,60
Each
Add to cart
Chemical Identifiers
CAS705-24-8
IUPAC Name4,6-dichloro-2-(trifluoromethyl)pyrimidine
Molecular FormulaC5HCl2F3N2
InChI KeyQFWVAJQVYBRTCL-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC(Cl)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only