Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%

CAS: 42521-10-8 | C8H8ClNO3 | 201.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H33438.06
also known as H33438-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS42521-10-8
IUPAC Namemethyl 2-chloro-6-methoxypyridine-4-carboxylate
Molecular FormulaC8H8ClNO3
InChI KeyDAPAXVAUEVRBGS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(OC)=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)98.0-106?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only