4-Acetamidobenzyl alcohol, 97%
4-Acetamidobenzyl alcohol, 97%
4-Acetamidobenzyl alcohol, 97%
Thermo Scientific Chemicals

4-Acetamidobenzyl alcohol, 97%

CAS: 16375-88-5 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33454.03
also known as H33454-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS16375-88-5
IUPAC NameN-[4-(hydroxymethyl)phenyl]acetamide
Molecular FormulaC9H11NO2
InChI KeyXEYORFKUJZEQCH-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)118.5-124.5?C
Appearance (Color)Pale yellow
Assay (HPLC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only