3-Amino-5-phenyl-1,2,4-thiadiazole, 96%
3-Amino-5-phenyl-1,2,4-thiadiazole, 96%
3-Amino-5-phenyl-1,2,4-thiadiazole, 96%
Thermo Scientific Chemicals

3-Amino-5-phenyl-1,2,4-thiadiazole, 96%

CAS: 27182-54-3 | C8H7N3S | 177.225 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H33552.03
also known as H33552-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS27182-54-3
IUPAC Name5-phenyl-1,2,4-thiadiazol-3-amine
Molecular FormulaC8H7N3S
InChI KeyWOZDQVLZCXNJPY-UHFFFAOYSA-N
SMILESNC1=NSC(=N1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)134.0-143.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only