Thermo Scientific Chemicals

4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H33610.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H33610.06
5 g, Each
Quantity
Catalog number: H33610.06
also known as H33610-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
189681-04-7
IUPAC Name
4-chloro-5-(thiophen-2-yl)thieno[2,3-d]pyrimidine
Molecular Formula
C10H5ClN2S2
InChI Key
VNWBUFWVNCUUKY-UHFFFAOYSA-N
SMILES
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
Appearance (Color)
White to pale yellow
Melting Point (clear melt)
108.0-117.0?C
Form
Powder
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text