Thermo Scientific Chemicals

Diethyl ether, Spectrophotometric Grade, 99+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: H33678.AU
1000 mL, Each
Thermo Scientific Chemicals

Diethyl ether, Spectrophotometric Grade, 99+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: H33678.AU
1000 mL, Each
Quantity
Catalog number: H33678.AU
also known as H33678-AU
Price (TWD)

Chemical Identifiers

CAS
60-29-7
IUPAC Name
ethoxyethane
Molecular Formula
C4H10O
InChI Key
RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES
CCOCC
Appearance (Color)
Clear, colourless
Assay (GC)
>99.0%
Residue on Evaporation
<1ppm
Form
liquid
Comment
Stabiliser: Copper gauze

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text