Thermo Scientific Chemicals

3-Fluoro-5-(pentafluorothio)aniline, 97%, Thermo Scientific Chemicals

Catalog number: H33680.MD
250 mg, Each
Thermo Scientific Chemicals

3-Fluoro-5-(pentafluorothio)aniline, 97%, Thermo Scientific Chemicals

Catalog number: H33680.MD
250 mg, Each
Quantity
Catalog number: H33680.MD
also known as H33680-MD
Price (USD)
Price: 188.00
Online price: 162.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1240256-99-8
IUPAC Name
3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Formula
C6H5F6NS
InChI Key
RFXWDLYDFAZNLO-UHFFFAOYSA-N
SMILES
NC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F
Form
Crystalline powder or fused solid
Melting Point (clear melt)
34-40?C
Assay (GC)
≥96.0%
Appearance (Color)
White to pale cream or pale grey

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text