Diethyl chlorophosphate, 97+%
Diethyl chlorophosphate, 97+%
Diethyl chlorophosphate, 97+%
Diethyl chlorophosphate, 97+%
Thermo Scientific Chemicals

Diethyl chlorophosphate, 97+%

CAS: 814-49-3 | C4H10ClO3P | 172.545 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H33820.22
also known as H33820-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS814-49-3
IUPAC Namediethyl phosphorochloridate
Molecular FormulaC4H10ClO3P
InChI KeyLGTLXDJOAJDFLR-UHFFFAOYSA-N
SMILESCCOP(Cl)(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.4140-1.4190 @ 20°C
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. James McNulty; Venkatesan Krishnamoorthy; Al Robertson. Direct formation of esters and amides from carboxylic acids using diethyl chlorophosphate in pyridine. Tetrahedron Letters. 200849 (44), 6344-6347
  2. Zarei, Maaroof. One-pot sequence synthesis of azetidin-2-one using diethyl chlorophosphate. Journal of Chemical Research. 201236 (2), 118-120(3)