Thermo Scientific Chemicals

5-Bromophthalide, 98%, Thermo Scientific Chemicals

Catalog number: H33891.22
100 g, Each
Thermo Scientific Chemicals

5-Bromophthalide, 98%, Thermo Scientific Chemicals

Catalog number: H33891.22
100 g, Each
Quantity
Catalog number: H33891.22
also known as H33891-22
Price (USD)
Price: 357.00
Online price: 308.65
Your price:
Quantity
-

Chemical Identifiers

CAS
64169-34-2
IUPAC Name
5-bromo-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C8H5BrO2
InChI Key
IUSPXLCLQIZFHL-UHFFFAOYSA-N
SMILES
BrC1=CC=C2C(=O)OCC2=C1
Assay (GC)
≥97.5% (non-U.S. sourced material)
Water Content (Karl Fischer Titration)
≤1.0% (non-U.S. sourced material)
Appearance (Color)
White to cream to brown or yellow to orange
Comment
Material Sourced in the U.S. and in other countries
Melting Point (clear melt)
161.0-168.0°C

Description

5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

Solubility
Soluble in acetone.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text