10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%
Thermo Scientific Chemicals

10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%

CAS: 944721-52-2 | C20H29F5O5Si | 472.524 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34117.MD
also known as H34117-MD
Price (EUR)
49,80
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
49,80
Each
Chemical Identifiers
CAS944721-52-2
IUPAC Name2,3,4,5,6-pentafluorophenyl 11-(trimethoxysilyl)undecanoate
Molecular FormulaC20H29F5O5Si
InChI KeyKSYPNQIBGIOYPZ-UHFFFAOYSA-N
SMILESCO[Si](CCCCCCCCCCC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4360-1.4420 @ 20?C
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
RUO – Research Use Only
Oriol Penon, et al. Optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.Colloids Surf., B.,2014,116(1), 104-113.