3-(9-Anthryl)propionic acid, 96%
3-(9-Anthryl)propionic acid, 96%
3-(9-Anthryl)propionic acid, 96%
Thermo Scientific Chemicals

3-(9-Anthryl)propionic acid, 96%

CAS: 41034-83-7 | C17H14O2 | 250.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34184.03
also known as H34184-03
Price (EUR)
102,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
102,00
Each
Chemical Identifiers
CAS41034-83-7
IUPAC Name3-(anthracen-9-yl)propanoic acid
Molecular FormulaC17H14O2
InChI KeyBDGMYCZUEIGHJH-UHFFFAOYSA-N
SMILESOC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream to yellow
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
Melting Point (clear melt)189.5-198.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only