2'-Fluoro-4-biphenylmethanol, 97%
2'-Fluoro-4-biphenylmethanol, 97%
2'-Fluoro-4-biphenylmethanol, 97%
Thermo Scientific Chemicals

2'-Fluoro-4-biphenylmethanol, 97%

CAS: 773872-57-4 | C13H11FO | 202.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34211.MD
also known as H34211-MD
Price (TWD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS773872-57-4
IUPAC Name{2'-fluoro-[1,1'-biphenyl]-4-yl}methanol
Molecular FormulaC13H11FO
InChI KeyWABWIEVUZWQZLC-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
Melting Point (clear melt)53.0-62.0?C
FormPowder or lumps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only