1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals
1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals
1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals
1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H34352.03
also known as H34352-03
1 g
Catalog number H34352.03
also known as H34352-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS243641-37-4
IUPAC Nametert-butyl 7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine-1-carboxylate
Molecular FormulaC14H20N2O2
InChI KeyCHWMFCCGEWQABK-UHFFFAOYSA-N
SMILESCC1=CC=C2CCCN(C(=O)OC(C)(C)C)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)93.0-102.0°C
FormCrystals or powder or crystalline powder
Appearance (Color)Pale yellow to yellow
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only