HCS LipidTOX™ Red Neutral Lipid Stain, for cellular imaging
Invitrogen™

HCS LipidTOX™ Red Neutral Lipid Stain, for cellular imaging

The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/orRead more
Have Questions?
Catalog NumberQuantity
H344761 each
Catalog number H34476
Price (KRW)
730,000
Online offer
Ends: 31-Dec-2026
811,000
Save 81,000 (10%)
Each
Quantity:
1 each
Price (KRW)
730,000
Online offer
Ends: 31-Dec-2026
811,000
Save 81,000 (10%)
Each
The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX™ Red neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX™ neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX™ phospholipidosis detection reagents (H34350, H34351). HCS LipidTOX™ neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use With (Equipment)High Content Instrument
Product LineLipidTOX
Quantity1 each
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeLipid Stain
Sub Cellular LocalizationCell Membranes and Lipids , Lipids
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

What kind of filter sets can I use with HCS LipidTOX neutral lipid stains?

LipidTOX Green neutral lipid stain can be imaged with filter sets appropriate for Alexa Fluor 488 dye or fluorescein. LipidTOX Red neutral lipid stain is best imaged with filter sets appropriate for Alexa Fluor 594 dye or Texas Red dye. LipidTOX Deep Red neutral lipid stain can imaged with filter sets appropriate for Alexa Fluor 647 dye or Cy5 dye.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (16)

Citations & References
Abstract
Vascular lipid accumulation, lipoprotein oxidation, and macrophage lipid uptake in hypercholesterolemic zebrafish.
Authors:Stoletov K, Fang L, Choi SH, Hartvigsen K, Hansen LF, Hall C, Pattison J, Juliano J, Miller ER, Almazan F, Crosier P, Witztum JL, Klemke RL, Miller YI,
Journal:Circ Res
PubMed ID:19265037
'Lipid accumulation in arteries induces vascular inflammation and atherosclerosis, the major cause of heart attack and stroke in humans. Extreme hyperlipidemia induced in mice and rabbits enables modeling many aspects of human atherosclerosis, but microscopic examination of plaques is possible only postmortem. Here we report that feeding adult zebrafish (Danio ... More
Enhancement of BODIPY505/515 lipid fluorescence method for applications in biofuel-directed microalgae production.
Authors:Brennan L, Blanco Fernández A, Mostaert AS, Owende P,
Journal:J Microbiol Methods
PubMed ID:22521923
'This paper describes a microalgal cell lipid fluorescence enhancement method using BODIPY(505/515), which can be used to screen for lipids in wild-type microalgae and to monitor lipid content within microalgae production processes to determine optimal harvesting time. The study was based on four microalgae species (Dunaliella teteriolecta, Tetraselmis suecica, Nannochloropsis ... More
Increased lipid accumulation in the Chlamydomonas reinhardtii sta7-10 starchless isoamylase mutant and increased carbohydrate synthesis in complemented strains.
Authors:Work VH, Radakovits R, Jinkerson RE, Meuser JE, Elliott LG, Vinyard DJ, Laurens LM, Dismukes GC, Posewitz MC,
Journal:Eukaryot Cell
PubMed ID:20562225
'The accumulation of bioenergy carriers was assessed in two starchless mutants of Chlamydomonas reinhardtii (the sta6 [ADP-glucose pyrophosphorylase] and sta7-10 [isoamylase] mutants), a control strain (CC124), and two complemented strains of the sta7-10 mutant. The results indicate that the genetic blockage of starch synthesis in the sta6 and sta7-10 mutants ... More
Knockdown of ACAT-1 reduces amyloidogenic processing of APP.
Authors:Huttunen HJ, Greco C, Kovacs DM
Journal:FEBS Lett
PubMed ID:17412327
'Previous studies have shown that acyl-coenzyme A:cholesterol acyl transferase (ACAT), an enzyme that controls cellular equilibrium between free cholesterol and cholesteryl esters, modulates proteolytic processing of APP in cell-based and animal models of Alzheimer''s disease. Here we report that ACAT-1 RNAi reduced cellular ACAT-1 protein by approximately 50% and cholesteryl ... More
Microplate-based high throughput screening procedure for the isolation of lipid-rich marine microalgae.
Authors:Pereira H, Barreira L, Mozes A, Florindo C, Polo C, Duarte CV, Custódio L, Varela J,
Journal:Biotechnol Biofuels
PubMed ID:22192119
We describe a new selection method based on BODIPY (4,4-difluoro-1,3,5,7-tetramethyl-4-bora-3a,4a-diaza-s-indacene) staining, fluorescence activated cell sorting (FACS) and microplate-based isolation of lipid-rich microalgae from an environmental sample. Our results show that direct sorting onto solid medium upon FACS can save about 3 weeks during the scale-up process as compared with the ... More