3-(2-Methoxyphenyl)-1H-pyrazole, 97%
3-(2-Methoxyphenyl)-1H-pyrazole, 97%
3-(2-Methoxyphenyl)-1H-pyrazole, 97%
3-(2-Methoxyphenyl)-1H-pyrazole, 97%
Thermo Scientific Chemicals

3-(2-Methoxyphenyl)-1H-pyrazole, 97%

CAS: 59843-63-9 | C10H10N2O | 174.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34492.03
also known as H34492-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS59843-63-9
IUPAC Name5-(2-methoxyphenyl)-1H-pyrazole
Molecular FormulaC10H10N2O
InChI KeyKLPGJCMICASHKV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥96.0%
Melting Point80.0-86.0°C
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only