6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%
Thermo Scientific Chemicals

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%

CAS: 138642-97-4 | C8H5ClN2O2 | 196.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H35567.03
also known as H35567-03
Price (EUR)
171,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
171,00
Each
Specifications
CAS138642-97-4
Chemical Name or Material6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate
Formula Weight196.59 (Anhydrous)
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only