6-Cyanoimidazo[1,2-a]pyridine, 95%
6-Cyanoimidazo[1,2-a]pyridine, 95%
6-Cyanoimidazo[1,2-a]pyridine, 95%
6-Cyanoimidazo[1,2-a]pyridine, 95%
Thermo Scientific Chemicals

6-Cyanoimidazo[1,2-a]pyridine, 95%

CAS: 106850-34-4 | C8H5N3 | 143.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H35854.03
also known as H35854-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS106850-34-4
IUPAC Nameimidazo[1,2-a]pyridine-6-carbonitrile
Molecular FormulaC8H5N3
InChI KeyLRJOKNYELSECDZ-UHFFFAOYSA-N
SMILESN#CC1=CN2C=CN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
FormPowder
Melting Point (clear melt)160.0-172.0?C
Appearance (Color)Cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only