n-Butyllithium, 1.6M in hexanes, packaged under Nitrogen in resealable AcroSeal™ bottles
n-Butyllithium, 1.6M in hexanes, packaged under Nitrogen in resealable AcroSeal™ bottles
n-Butyllithium, 1.6M in hexanes, packaged under Nitrogen in resealable AcroSeal™ bottles
Thermo Scientific Chemicals

n-Butyllithium, 1.6M in hexanes, packaged under Nitrogen in resealable AcroSeal™ bottles

CAS: 109-72-8 | C{4}H{9}Li
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
800 mL
產品號碼 H36033.AP
亦稱為 H36033-AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
檢視更多
規格Specification Sheet規格表
Assay from Supplier's CofA≥14.0 to ≤16.0% (Active Base Titration)
FormLiquid
Free base (titration)≤1.0%
Concentration1.49 to 1.70M
Appearance (Color)Clear to turbid colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.