Diloxanide (furoate), 97%
Diloxanide (furoate), 97%
Diloxanide (furoate), 97%
Thermo Scientific™

Diloxanide (furoate), 97%

Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
產品號碼 H36096.MD
亦稱為 H36096-MD
價格 (TWD)
-
Quantity:
250 mg
Request bulk or custom format
規格
CAS3736-81-0
EINECS Number223-108-1
Formula Weight328.15
IUPAC Name4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate
InChI KeyBDYYDXJSHYEDGB-UHFFFAOYSA-N
檢視更多