Radglo GM-10 Chartreuse, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Radglo GM-10 Chartreuse, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H36123.22
also known as H36123-22
100 g
Catalog number H36123.22
also known as H36123-22
Price (EUR)
45,40
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
45,40
Each
Specifications
Chemical Name or MaterialRadglo GM-10 Chartreuse
Density1.3
Recommended StorageAmbient temperatures
TSCANo
Quantity100 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only