3-Yodo-1H-pirazol, 97 %, Thermo Scientific Chemicals
3-Yodo-1H-pirazol, 97 %, Thermo Scientific Chemicals
3-Yodo-1H-pirazol, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Yodo-1H-pirazol, 97 %, Thermo Scientific Chemicals

CAS: 4522-35-4 | C3H3IN2 | 193.98 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo H36465.06
también denominado H36465-06
Precio (EUR)
186,00
Each
Cantidad:
5 g
Pedido a granel o personalizado
Precio (EUR)
186,00
Each
Identificadores químicos
CAS4522-35-4
IUPAC Name5-iodo-1H-pyrazole
Molecular FormulaC3H3IN2
InChI KeyRUKDVLFJSMVBLV-UHFFFAOYSA-N
SMILESIC1=CC=NN1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormPowder
Appearance (Color)Yellow
Assay (GC)≥96.0%
Melting Point (clear melt)66.0-75.0?C
3-Iodo-1H-pyrazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 3-yodo-1H-pirazol se utiliza como intermedio farmacéutico.

Solubilidad
Insoluble en agua.

Notas
Sensible a la luz. Incompatible con agentes oxidantes, ácidos y bases.
RUO – Research Use Only

General References:

  1. Rita Mazeikaite; Jurgis Sudzius; Gintaras Urbelis; Linas Labanauskas. Synthesis of substituted-3-iodo-1H-pyrazole derivatives and their further modification under Sonogashira cross-coupling reaction conditions. ARKIVOC. 2014, 6, 54-71
  2. Taiki Morita; Daisuke Kobayashi; Keisuke Matsumura; Dr. Kohei Johmoto; Dr. Hidehiro Uekusa; Dr. Shinichiro Fuse; Prof. Dr. Takashi Takahash. Sequential SNAr reaction/Suzuki-Miyaura Coupling/C-H direct arylations approach for the rapid synthesis of tetraaryl-substituted pyrazoles. Chemistry - An Asian Journal. 2015, 10, (8),1626-1630