tert-Butyllithium, nominally 1.9M in pentane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
tert-Butyllithium, nominally 1.9M in pentane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyllithium, nominally 1.9M in pentane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H36881.AE
also known as H36881-AE
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
Add to cart
tert-Butyllithium, nominally 1.9M in pentane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Catalog numberH36881.AE
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametert-butyllithium
Molecular FormulaC4H9Li
InChI KeyBKDLGMUIXWPYGD-UHFFFAOYSA-N
SMILES[Li]C(C)(C)C
Molecular Weight (g/mol)64.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to turbid colorless to yellow
FormLiquid
Assay from Supplier's CofA≥17.0 to ≤19.0% (Active Base Titration)
Concentration1.80 to 2.02M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.