Appearance (Color)Clear to turbid colorless to yellow
Concentration2.37 to 2.59M
Assay from Supplier's CofA≥22.0 to ≤24.0% (Active Base Titration)
Free base (titration)≤1.2%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.