Diisobutylaluminum hydride, 1.1M in cyclohexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diisobutylaluminum hydride, 1.1M in cyclohexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H37108.AE
also known as H37108-AE
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS1191-15-7
IUPAC Namebis(2-methylpropyl)alumanyl
Molecular FormulaC8H18Al
InChI KeySIPUZPBQZHNSDW-UHFFFAOYSA-N
SMILESCC(C)C[Al]CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥19.5 to 20.5% (DIBAL-H, on Al basis)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.