Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals
Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals
Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H37697.06
also known as H37697-06
5 g
Catalog number H37697.06
also known as H37697-06
Price (USD)
63.65
Online Exclusive
70.80
Save 7.15 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.80
Save 7.15 (10%)
Each
Chemical Identifiers
CAS4844-38-6
IUPAC Namediethyl dodecylphosphonate
Molecular FormulaC16H35O3P
InChI KeyLRIWHHSVZCKNJL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.0%
Appearance (Color)Clear colorless
Refractive Index1.4385-1.4435 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only