Methyl blue
Methyl blue
Methyl blue
Thermo Scientific Chemicals

Methyl blue

CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H37721.22
Price (EUR)
36,40
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
36,40
Each
Chemical Identifiers
CAS28983-56-4
IUPAC Namedisodium 2-{[4-({4-[(2-sulfonatophenyl)amino]phenyl}({4-[(2-sulfonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene})methyl)phenyl]amino}benzene-1-sulfonate
Molecular FormulaC37H26N3Na2O9S3
InChI KeyTUHAIJABPUJAEY-UHFFFAOYSA-K
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to dark brown
UV/Vis≥600 (@ lambda-max, in Water: pH 3)
FormPowder or crystalline powder
lambda-max (UV-Vis)595-605nm (in Water: pH 3)
Loss on Drying≤10.0% (@ 200°C)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only