Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50157.03
also known as H50157-03
1 g
Catalog number H50157.03
also known as H50157-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS902837-55-2
Chemical Name or MaterialEthyl 2-chloro-5-pyridylglyoxylate
Formula Weight213.62
IUPAC Nameethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
InChI KeyHRFPVENOVYSKHE-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only