1-Phenylcyclohexanecarboxylic acid, 99%
1-Phenylcyclohexanecarboxylic acid, 99%
1-Phenylcyclohexanecarboxylic acid, 99%
Thermo Scientific Chemicals

1-Phenylcyclohexanecarboxylic acid, 99%

CAS: 1135-67-7 | C13H16O2 | 204.269 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50203.06
also known as H50203-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS1135-67-7
Chemical Name or Material1-Phenylcyclohexanecarboxylic acid
EINECS Number214-495-8
Formula Weight204.27
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only