4-(Methylsulfonyl)benzoic acid, 98%
4-(Methylsulfonyl)benzoic acid, 98%
4-(Methylsulfonyl)benzoic acid, 98%
Thermo Scientific Chemicals

4-(Methylsulfonyl)benzoic acid, 98%

CAS: 4052-30-6 | C8H8O4S | 200.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50230.06
also known as H50230-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4052-30-6
IUPAC Name4-methanesulfonylbenzoic acid
Molecular FormulaC8H8O4S
InChI KeyAJBWNNKDUMXZLM-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
Loss on Drying≤1%
Appearance (Color)White
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only