2-Cyanobenzoic acid, 94%
2-Cyanobenzoic acid, 94%
2-Cyanobenzoic acid, 94%
Thermo Scientific Chemicals

2-Cyanobenzoic acid, 94%

CAS: 3839-22-3 | C8H5NO2 | 147.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50237.03
also known as H50237-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3839-22-3
IUPAC Name2-cyanobenzoate
Molecular FormulaC8H4NO2
InChI KeyDTNSDCJFTHMDAK-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale grey
FormCrystals or powder or crystalline powder
Assay (HPLC)≥92.0%
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Anlai Wang; John A.Maguire; Ed Biehl. Preparation of 2-cyanobenzoic acids from the reaction of bromobenzoic acids with arylacetonitriles and LDA. J. Org. Chem. 1998, 63, (8),2451-2455
  2. Dacio Adhemar Iglesias; Danila Lujan Ruiz; Patricia Ercilia Allegretti. Experimental study of tautomeric equilibria of 2-cyanobenzoic acids in gas, solution and solid phase. Journal of Applied Solution Chemistry and Modeling. 2012, 1, 113-120