Specifications
CAS254732-51-9
Chemical Name or Material2-Chloro-3-(trifluoromethyl)quinoxaline
Formula Weight232.59
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more