3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%
3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%
3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%
Thermo Scientific Chemicals

3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%

CAS: 163295-70-3 | C7H5Cl3O2S | 259.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50707.03
also known as H50707-03
Price (USD)
152.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
152.00
Each
Specifications
CAS163295-70-3
Chemical Name or Material3,5-Dichloro-alpha-toluenesulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight259.54
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only