1-Aza-15-corona-5, 97 %, Thermo Scientific Chemicals
1-Aza-15-corona-5, 97 %, Thermo Scientific Chemicals
1-Aza-15-corona-5, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Aza-15-corona-5, 97 %, Thermo Scientific Chemicals

CAS: 66943-05-3 | C10H21NO4 | 219.281 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 mg
1 g
5 g
Número de catálogo H51064.03
también denominado H51064-03
Precio (EUR)
241,00
Each
Cantidad:
1 g
Pedido a granel o personalizado
Precio (EUR)
241,00
Each
Identificadores químicos
CAS66943-05-3
IUPAC Name1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Molecular FormulaC10H22NO4
InChI KeyBJUOQSZSDIHZNP-UHFFFAOYSA-O
SMILESC1COCCOCCOCCOCC[NH2+]1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
CommentConverts to solid at low temperature
Appearance (Color)Pale yellow
Assay by NMR>96.0%
FormOil
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La 1-Aza-15-corona-5 se utiliza como compuesto básico en la síntesis de los éteres bis-corona, los éteres lariat y otros macrociclos monofuncionalizados. También se utiliza como intermedio farmacéutico.

Solubilidad
Soluble en agua y metanol.

Notas
Sensible al aire. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only

General References:

  1. Pu Qian; Toshiro Yokoyama; Osamu Itabashi; Tomio Goto; Tokuji Miyashita. Ag+ ion sensing by the polymer Langmuir-Blodgett films containing 1-aza-15-crown 5-ether group. Thin Solid Films. 1997, 305, (1-2),316-320
  2. Kh. A.Hassan. Spectroscopic studies of complexes of iodine with the bases 1-aza-15-crown-5 and 3,6,9,14-tetrathiabicyclo[9.2.1]tetradeca-11,13-diene. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2004, 60, (13),3059-3063