4-(Tri-n-butylstannyl)thiazole, 95%
4-(Tri-n-butylstannyl)thiazole, 95%
4-(Tri-n-butylstannyl)thiazole, 95%
Thermo Scientific Chemicals

4-(Tri-n-butylstannyl)thiazole, 95%

CAS: 173979-01-6 | C15H29NSSn | 374.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51457.03
also known as H51457-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS173979-01-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥94.0%
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Rudiger Wittenberg; Jiri Srogl; Masahiro Egi; Lanny S.Liebeskind. Ketone synthesis under neutral conditions. Cu(I) diphenylphosphinate-mediated, palladium-catalyzed coupling of thiol esters and organostannanes. Org. Lett. 2003, 5, (17),3033-3035
  2. Hao Li; Hao Yang; Lanny S.Liebeskind. Synthesis of high enantiopurity N-Protected α-amino ketones by thiol ester-Organostannane cross-coupling using pH-neutral conditions. Org. Lett. 2008, 10, (19),4375-4378