N-Fmoc-4-nitro-L-phenylalanine, 98%
N-Fmoc-4-nitro-L-phenylalanine, 98%
N-Fmoc-4-nitro-L-phenylalanine, 98%
N-Fmoc-4-nitro-L-phenylalanine, 98%
Thermo Scientific Chemicals

N-Fmoc-4-nitro-L-phenylalanine, 98%

CAS: 95753-55-2 | C24H20N2O6 | 432.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51962.06
also known as H51962-06
Price (EUR)
148,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
148,00
Each
Add to cart
Chemical Identifiers
CAS95753-55-2
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid
Molecular FormulaC24H20N2O6
InChI KeyRZRRJPNDKJOLHI-QFIPXVFZSA-N
SMILESOC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Optical Rotation|b = -34? ?1? (C=1.055 in DMF)
Appearance (Color)White to pale cream
FormFused solid
Melting Point179 - 181?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only