N-Boc-3-chloro-L-phenylalanine, 95%
N-Boc-3-chloro-L-phenylalanine, 95%
N-Boc-3-chloro-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Boc-3-chloro-L-phenylalanine, 95%

CAS: 114873-03-9 | C14H18ClNO4 | 299.751 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52015.06
also known as H52015-06
Price (EUR)
310,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
310,00
Each
Specifications
CAS114873-03-9
Chemical Name or MaterialN-Boc-3-chloro-L-phenylalanine
Formula Weight299.75
IUPAC Name(2S)-3-(3-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
InChI KeyRCZHBTHQISEPPP-NSHDSACASA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only